siRNA Details
Gene Symbol: WDR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242418.1 | NP_001229347.1 | 2, 3 | 545 | ||
NM_144574.3 | NP_653175.2 | 2, 3 | 509 | |||
NM_181291.2 | NP_851808.1 | 2, 3 | 509 | |||
XM_006720309.1 | XP_006720372.1 | 2, 3 | 292 | |||
XM_006720310.1 | XP_006720373.1 | 2, 3 | 253 | |||
XM_011537341.2 | XP_011535643.1 | 2, 3 | 527 | |||
XM_011537344.2 | XP_011535646.1 | 3, 4 | 514 | |||
XM_011537346.1 | XP_011535648.1 | 2, 3 | 318 | |||
XM_017021767.1 | XP_016877256.1 | 2, 3 | 527 | |||
XM_017021768.1 | XP_016877257.1 | 2, 3 | 542 | |||
XM_017021770.1 | XP_016877259.1 | 2, 3 | 542 | |||
XM_017021771.1 | XP_016877260.1 | 3, 4 | 577 | |||
XM_017021772.1 | XP_016877261.1 | 3, 4 | 330 | |||
XM_017021775.1 | XP_016877264.1 | 3, 4 | 378 | |||
XM_017021776.1 | XP_016877265.1 | 2, 3 | 292 | |||
XM_017021777.1 | XP_016877266.1 | 2, 3 | 260 | |||
GenBank mRNA | AF327354.1 | - | 2, 3 | 434 | ||
AK056330.1 | - | 2, 3 | 251 | |||
AL832500.1 | - | 2, 3 | 484 | |||
BC028387.2 | - | 2, 3 | 468 | 4823662 | ||
BP363522.1 | - | 2, 3 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
