siRNA Details
Gene Symbol: SLC39A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213568.1 | NP_998733.1 | 2 | 522 | ||
GenBank mRNA | AK002044.1 | - | 2 | 485 | ||
BC020571.2 | - | 2 | 490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
