siRNA Details
Gene Symbol: NIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142275.1 | NP_001135747.1 | 4 | 313 | ||
NM_144599.4 | NP_653200.2 | 4 | 492 | |||
GenBank mRNA | AK057846.1 | - | 4 | 347 | ||
AK314073.1 | - | 4 | 453 | |||
BC025678.1 | - | 4 | 313 | |||
BC103705.1 | - | 5 | 439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
