siRNA Details
Gene Symbol: ZNF513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056765.1 | - | 2 | 1321 | ||
AK096933.1 | - | 3 | 1878 | |||
BC040650.1 | - | 2 | 1310 | |||
BC052282.1 | - | 4 | 1998 | 5467936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
