siRNA Details
Gene Symbol: DOCK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144658.3 | NP_653259.3 | 34 | 3813 | ||
XM_005262368.3 | XP_005262425.1 | 35 | 4097 | |||
XM_005262369.4 | XP_005262426.1 | 35 | 3791 | |||
XM_011531276.2 | XP_011529578.1 | 35 | 3732 | |||
XM_011531278.2 | XP_011529580.1 | 22 | 2218 | |||
GenBank mRNA | AK056684.1 | - | 4 | 327 | ||
AK124735.1 | - | 29 | 3236 | |||
AK125641.1 | - | 13 | 1325 | |||
AY692226.1 | - | 34 | 3813 |
