siRNA Details
Gene Symbol: PSMA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025096.1 | NP_001020267.1 | 2 | 237 | ||
NM_001308188.1 | NP_001295117.1 | 3 | 307 | |||
NM_144662.2 | NP_653263.2 | 2 | 237 | |||
XM_011525818.1 | XP_011524120.1 | 3 | 172 | |||
GenBank mRNA | BC021713.1 | - | 3 | 247 | 4826048 | |
BC025389.1 | - | 2 | 215 | |||
BC042820.1 | - | 2 | 237 | 5268996 | ||
BC047355.1 | - | 2 | 211 | |||
BI461560.1 | - | 2 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
