siRNA Details
Gene Symbol: DUOXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276267.1 | NP_001263196.1 | 9 | 2324 | ||
NM_001276268.1 | NP_001263197.1 | 6 | 1984 | |||
XM_006720743.3 | XP_006720806.1 | 9 | 2654 | |||
XM_006720745.1 | XP_006720808.1 | 9 | 2629 | |||
XM_006720746.1 | XP_006720809.1 | 9 | 2658 | |||
XM_006720747.3 | XP_006720810.1 | 8 | 2566 | |||
XM_006720749.1 | XP_006720812.1 | 9 | 2424 | |||
XM_006720751.3 | XP_006720814.1 | 7 | 2147 | |||
XM_006720752.3 | XP_006720815.2 | 7 | 2156 | |||
XM_006720754.1 | XP_006720817.1 | 8 | 2289 | |||
XM_006720755.1 | XP_006720818.1 | 8 | 2265 | |||
XM_011522181.2 | XP_011520483.1 | 9 | 2682 | |||
XM_011522182.1 | XP_011520484.1 | 8 | 2397 | |||
XM_011522183.2 | XP_011520485.1 | 10 | 2512 | |||
GenBank mRNA | AK056896.1 | - | 6 | 1984 |
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM987789.1 | - | 1 | 46 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
