siRNA Details
Gene Symbol: TAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270488.1 | NP_001257417.1 | 2 | 334 | ||
NM_005643.3 | NP_005634.1 | 2 | 334 | |||
XM_011514827.2 | XP_011513129.1 | 2 | 724 | |||
GenBank mRNA | AF118094.1 | - | 2 | 74 | ||
AK302226.1 | - | 2 | 305 | |||
AK313475.1 | - | 2 | 275 | |||
BC021972.2 | - | 2 | 290 | 3935882 | ||
BT019404.1 | - | 2 | 220 | |||
BT019405.1 | - | 2 | 220 | |||
BX538195.1 | - | 1 | 131 | |||
BX647568.1 | - | 1 | 502 | |||
CR542068.1 | - | 2 | 220 | |||
D63705.1 | - | 2 | 303 | |||
DA153455.1 | - | 2 | 334 | |||
X83928.1 | - | 2 | 312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
