siRNA Details
Gene Symbol: CCDC155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144688.4 | NP_653289.3 | 3 | 312 | ||
XM_005258549.3 | XP_005258606.1 | 3 | 312 | |||
XM_011526488.2 | XP_011524790.1 | 3 | 312 | |||
XM_011526489.2 | XP_011524791.1 | 3 | 2212 | |||
XM_011526490.2 | XP_011524792.1 | 3 | 312 | |||
XM_011526494.2 | XP_011524796.1 | 4 | 285 | |||
XM_011526495.2 | XP_011524797.1 | 4 | 321 | |||
XM_011526496.2 | XP_011524798.1 | 2 | 173 | |||
XM_011526497.2 | XP_011524799.1 | 3 | 271 | |||
XM_017026333.1 | XP_016881822.1 | 2 | 445 | |||
GenBank mRNA | AK057220.1 | - | 3 | 281 | ||
AK097696.1 | - | 4 | 1025 | |||
BC029811.1 | - | 3 | 231 | 5171712 | ||
DB024870.1 | - | 3 | 312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
