siRNA Details
Gene Symbol: ZNF582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320371.1 | NP_001307300.1 | 3 | 553 | ||
NM_144690.2 | NP_653291.1 | 3 | 331 | |||
GenBank mRNA | AK055489.1 | - | 3 | 250 | ||
AK299036.1 | - | 3 | 477 | |||
AK316028.1 | - | 3 | 497 | |||
BC011796.2 | - | 3 | 281 | |||
BC056238.1 | - | 3 | 268 | |||
BC101008.1 | - | 2 | 172 | 40006516 | ||
BC101009.1 | - | 2 | 172 | 40006517 | ||
BC101010.1 | - | 2 | 172 | 40006519 | ||
BC101011.1 | - | 2 | 173 | 40006520 | ||
DA929494.1 | - | 3 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
