siRNA Details
Gene Symbol: AXDND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144696.5 | NP_653297.3 | 21, 22 | 2891 | ||
NR_073544.1 | 21, 22 | 2624 | ||||
XM_011509165.1 | XP_011507467.1 | 21, 22 | 2870 | |||
XM_011509166.2 | XP_011507468.1 | 21, 22 | 2917 | |||
XM_011509167.2 | XP_011507469.1 | 21, 22 | 3032 | |||
XM_011509168.1 | XP_011507470.1 | 21, 22 | 2874 | |||
XM_011509169.1 | XP_011507471.1 | 20, 21 | 2663 | |||
XM_011509171.1 | XP_011507473.1 | 20, 21 | 2688 | |||
XM_011509176.1 | XP_011507478.1 | 19, 20 | 2573 | |||
XM_011509179.2 | XP_011507481.1 | 16, 17 | 1978 | |||
XM_011509181.2 | XP_011507483.1 | 11, 12 | 1487 | |||
XM_017000257.1 | XP_016855746.1 | 15, 16 | 1838 | |||
XM_017000258.1 | XP_016855747.1 | 15, 16 | 1870 | |||
XM_017000259.1 | XP_016855748.1 | 7, 8 | 1040 | |||
GenBank mRNA | AK058167.1 | - | 3, 4 | 248 | ||
BC146823.1 | - | 21, 22 | 2625 | 8860438 | ||
BX647935.1 | - | 21, 22 | 2893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
