siRNA Details
Gene Symbol: CDKL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037343.1 | NP_001032420.1 | 3, 4 | 321 | ||
NM_001323289.1 | NP_001310218.1 | 1, 2 | 234 | |||
NM_003159.2 | NP_003150.1 | 2, 3 | 325 | |||
GenBank mRNA | AK307444.1 | - | 2, 3 | 322 | ||
AL704691.1 | - | 3, 4 | 360 | |||
AY217744.1 | - | 2, 3 | 293 | |||
HQ171445.1 | - | 1, 2 | 72 | |||
Y15057.1 | - | 2, 3 | 293 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001024624, BY291331, AK139220, AK052380.
Gene Ontology Categories:
