siRNA Details
Gene Symbol: CCDC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001158261.1 | NP_001151733.1 | 3 | 481 | ||
NM_144681.2 | NP_653282.2 | 3 | 481 | |||
GenBank mRNA | AK057296.1 | - | 3 | 481 | ||
BC029224.1 | - | 3 | 432 | 5172741 | ||
CV030241.1 | - | 3 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
