siRNA Details
Gene Symbol: PHOSPHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008489.3 | NP_001008489.1 | 4 | 636 | ||
NM_001199285.1 | NP_001186214.1 | 4 | 536 | |||
NM_001199286.1 | NP_001186215.1 | 4 | 755 | |||
NM_001199287.1 | NP_001186216.1 | 5 | 754 | |||
NM_001199288.1 | NP_001186217.1 | 4 | 555 | |||
GenBank mRNA | AK314915.1 | - | 4 | 539 | ||
BC022324.2 | - | 3 | 467 | 4701753 | ||
BC106013.1 | - | 4 | 625 | 4387304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
