siRNA Details
Gene Symbol: TEPSIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144679.2 | NP_653280.1 | 12 | 1951 | ||
XM_005257066.2 | XP_005257123.1 | 14 | 2749 | |||
XM_005257067.2 | XP_005257124.1 | 13 | 2155 | |||
XM_006721709.3 | XP_006721772.1 | 13 | 2162 | |||
XM_006721712.2 | XP_006721775.1 | 13 | 2593 | |||
XM_011524355.1 | XP_011522657.1 | 14 | 2755 | |||
XM_011524356.1 | XP_011522658.1 | 13 | 2551 | |||
XM_011524357.1 | XP_011522659.1 | 13 | 2783 | |||
XM_011524358.2 | XP_011522660.1 | 14 | 2684 | |||
XM_017024202.1 | XP_016879691.1 | 14 | 2996 | |||
XM_017024203.1 | XP_016879692.1 | 15 | 2643 | |||
XM_017024204.1 | XP_016879693.1 | 14 | 3196 | |||
GenBank mRNA | AK056090.1 | - | 12 | 1915 | ||
AK127221.1 | - | 11 | 3308 | |||
AK128728.1 | - | 14 | 3468 | |||
BC064483.1 | - | 12 | 1912 | 5430837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
