siRNA Details
Gene Symbol: AXDND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144696.5 | NP_653297.3 | 22 | 2978 | ||
NR_073544.1 | 22 | 2711 | ||||
XM_011509165.1 | XP_011507467.1 | 22 | 2957 | |||
XM_011509166.2 | XP_011507468.1 | 22 | 3004 | |||
XM_011509167.2 | XP_011507469.1 | 22 | 3119 | |||
XM_011509168.1 | XP_011507470.1 | 22 | 2961 | |||
XM_011509169.1 | XP_011507471.1 | 21 | 2750 | |||
XM_011509170.1 | XP_011507472.1 | 21 | 2849 | |||
XM_011509171.1 | XP_011507473.1 | 21 | 2775 | |||
XM_011509176.1 | XP_011507478.1 | 20 | 2660 | |||
XM_011509179.2 | XP_011507481.1 | 17 | 2065 | |||
XM_011509181.2 | XP_011507483.1 | 12 | 1574 | |||
XM_017000257.1 | XP_016855746.1 | 16 | 1925 | |||
XM_017000258.1 | XP_016855747.1 | 16 | 1957 | |||
XM_017000259.1 | XP_016855748.1 | 8 | 1127 | |||
GenBank mRNA | AK058167.1 | - | 4 | 335 | ||
BC146823.1 | - | 22 | 2712 | 8860438 | ||
BX647935.1 | - | 22 | 2980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
