siRNA Details
Gene Symbol: TTC23L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317949.1 | NP_001304878.1 | 11 | 1263 | ||
NM_144725.3 | NP_653326.3 | 11 | 1328 | |||
XM_011513984.2 | XP_011512286.1 | 12 | 1348 | |||
XM_017009119.1 | XP_016864608.1 | 12 | 1592 | |||
XM_017009120.1 | XP_016864609.1 | 12 | 1411 | |||
XM_017009121.1 | XP_016864610.1 | 12 | 1456 | |||
XM_017009122.1 | XP_016864611.1 | 12 | 1527 | |||
XM_017009124.1 | XP_016864613.1 | 10 | 1288 | |||
XM_017009125.1 | XP_016864614.1 | 10 | 1089 | |||
XM_017009126.1 | XP_016864615.1 | 10 | 1222 | |||
GenBank mRNA | AK058168.1 | - | 11 | 1328 | ||
BC057775.1 | - | 11 | 1300 | 5295348 | ||
BX088767.1 | - | 6 | 605 | |||
JN799997.1 | - | 11 | 1263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
