siRNA Details
Gene Symbol: RIBC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144968.3 | NP_659405.1 | 5 | 1688 | ||
GenBank mRNA | AK057345.1 | - | 5 | 1654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
