siRNA Details
Gene Symbol: RBMY2FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002193.2 | 4 | 1090 | |||
GenBank mRNA | BC026077.2 | - | 4 | 1092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
