siRNA Details
Gene Symbol: SHANK2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046567.1 | 3 | 1658 | |||
GenBank mRNA | AK097446.1 | - | 3 | 1656 |
Gene Symbol: ZNF564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144976.3 | NP_659413.1 | 4 | 2678 | ||
GenBank mRNA | BC028367.1 | - | 4 | 2615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
