siRNA Details
Gene Symbol: CEP112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199165.2 | NP_001186094.1 | 13 | 1443 | ||
NM_001302891.1 | NP_001289820.1 | 11 | 1393 | |||
XM_005257119.4 | XP_005257176.1 | 13 | 1537 | |||
XM_005257125.3 | XP_005257182.1 | 5 | 527 | |||
XM_005257126.4 | XP_005257183.1 | 2 | 192 | |||
XM_006721740.2 | XP_006721803.1 | 13 | 1409 | |||
XM_006721744.3 | XP_006721807.1 | 13 | 1541 | |||
XM_011524461.2 | XP_011522763.1 | 13 | 1550 | |||
XM_011524462.2 | XP_011522764.1 | 13 | 1539 | |||
XM_011524463.2 | XP_011522765.1 | 13 | 1539 | |||
XM_011524464.2 | XP_011522766.1 | 13 | 1539 | |||
XM_011524465.2 | XP_011522767.1 | 10 | 866 | |||
XM_011524466.2 | XP_011522768.1 | 3 | 345 | |||
XM_011524467.2 | XP_011522769.1 | 2 | 187 | |||
XM_017024302.1 | XP_016879791.1 | 11 | 1280 | |||
XM_017024303.1 | XP_016879792.1 | 13 | 1541 | |||
GenBank mRNA | AF458591.1 | - | 3 | 350 | ||
AK096929.1 | - | 13 | 1406 | |||
AK098070.1 | - | 5 | 475 | |||
AK299075.1 | - | 13 | 1394 | |||
BC037991.2 | - | 13 | 1505 | |||
BC131805.1 | - | 11 | 1221 | 40119069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
