siRNA Details
Gene Symbol: ZNF501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258280.1 | NP_001245209.1 | 3 | 2951 | ||
NM_145044.3 | NP_659481.2 | 3 | 2658 | |||
GenBank mRNA | AK054613.1 | - | 4 | 2486 | ||
BC013762.1 | - | 3 | 2634 | 4521303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
