siRNA Details
Gene Symbol: WDR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012361.3 | NP_001012361.1 | 4, 5 | 551 | ||
NM_145241.4 | NP_660284.1 | 4, 5 | 548 | |||
NR_134677.1 | 4, 5 | 480 | ||||
GenBank mRNA | AK093240.1 | - | 4, 5 | 480 | ||
AK313843.1 | - | 4, 5 | 490 | |||
AL833635.1 | - | 4, 5 | 397 | |||
BC012352.1 | - | 4, 5 | 480 | 4634445 | ||
BC022982.1 | - | 4, 5 | 477 | |||
DA209351.1 | - | 551 | ||||
DA305733.1 | - | 4, 5 | 471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
