siRNA Details
Gene Symbol: GLYATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145016.3 | NP_659453.3 | 5, 6 | 616 | ||
XM_011544809.2 | XP_011543111.1 | 5, 6 | 801 | |||
XM_017017337.1 | XP_016872826.1 | 6, 7 | 751 | |||
XM_017017338.1 | XP_016872827.1 | 5, 6 | 1198 | |||
GenBank mRNA | AB207211.1 | - | 4, 5 | 485 | ||
BC021682.1 | - | 5, 6 | 603 | 4079236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
