siRNA Details
Gene Symbol: OSCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145047.4 | NP_659484.4 | 10 | 1445 | ||
XM_005270461.1 | XP_005270518.1 | 11 | 1475 | |||
XM_005270462.1 | XP_005270519.1 | 10 | 1335 | |||
XM_011540680.2 | XP_011538982.1 | 12 | 1517 | |||
XM_011540681.2 | XP_011538983.1 | 11 | 1368 | |||
XM_017000303.1 | XP_016855792.1 | 9 | 1241 | |||
GenBank mRNA | AF462348.1 | - | 10 | 1487 | ||
AK098541.1 | - | 10 | 1291 | |||
BC018069.1 | - | 10 | 1411 | 4816622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
