siRNA Details
Gene Symbol: DNAJC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190233.1 | NP_001177162.1 | 4 | 346 | ||
NM_145261.3 | NP_660304.1 | 4 | 334 | |||
NR_033721.1 | 3 | 282 | ||||
GenBank mRNA | AI268262.1 | - | 4 | 333 | ||
AK309019.1 | - | 2 | 1235 | |||
AK311765.1 | - | 4 | 262 | |||
BC009702.1 | - | 4 | 286 | 3892297 | ||
BC073989.1 | - | 4 | 279 | 5760865 | ||
BE773017.1 | - | 4 | 301 | |||
BF666599.1 | - | 4 | 296 | |||
BG492744.1 | - | 3 | 218 | |||
CB054787.1 | - | 1 | 50 | |||
CB159371.1 | - | 4 | 346 | |||
KU178861.1 | - | 3 | 162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
