siRNA Details
Gene Symbol: AASDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286669.1 | NP_001273598.1 | 2 | 366 | ||
NM_001286670.1 | NP_001273599.1 | 2 | 366 | |||
NM_001286671.1 | NP_001273600.1 | 2 | 366 | |||
NM_001286672.1 | NP_001273601.1 | 2 | 366 | |||
NM_001323890.1 | NP_001310819.1 | 2 | 366 | |||
NM_001323892.1 | NP_001310821.1 | 2 | 366 | |||
NM_001323893.1 | NP_001310822.1 | 2 | 366 | |||
NM_001323899.1 | NP_001310828.1 | 2 | 366 | |||
NM_181806.3 | NP_861522.2 | 2 | 366 | |||
XM_017007740.1 | XP_016863229.1 | 2 | 327 | |||
XM_017007741.1 | XP_016863230.1 | 2 | 327 | |||
XM_017007743.1 | XP_016863232.1 | 2 | 327 | |||
GenBank mRNA | AK294332.1 | - | 2 | 328 | ||
AK303733.1 | - | 2 | 319 | |||
AK304309.1 | - | 2 | 357 | |||
AY314787.1 | - | 2 | 329 | |||
AY422212.1 | - | 2 | 281 | |||
BC015096.1 | - | 2 | 330 | |||
BC141824.1 | - | 2 | 324 | 40148174 | ||
BC142709.1 | - | 2 | 302 | 40148820 | ||
BX648853.1 | - | 2 | 358 | |||
DC306784.1 | - | 2 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
