siRNA Details
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 37, 38 | 6228 | ||
XM_006711956.2 | XP_006712019.1 | 37, 38 | 6333 | |||
XM_011532649.2 | XP_011530951.1 | 37, 38 | 6182 | |||
XM_011532650.2 | XP_011530952.1 | 37, 38 | 6183 | |||
XM_011532652.1 | XP_011530954.1 | 36, 37 | 6102 | |||
XM_011532653.1 | XP_011530955.1 | 37, 38 | 6228 | |||
XM_011532654.1 | XP_011530956.1 | 36, 37 | 5961 | |||
XM_011532655.2 | XP_011530957.1 | 33, 34 | 5250 | |||
XM_011532657.1 | XP_011530959.1 | 31, 32 | 4989 | |||
XM_011532660.1 | XP_011530962.1 | 37, 38 | 6228 | |||
XM_017003521.1 | XP_016859010.1 | 36, 37 | 6102 | |||
XM_017003522.1 | XP_016859011.1 | 32, 33 | 6054 | |||
XM_017003523.1 | XP_016859012.1 | 32, 33 | 5221 | |||
XM_017003524.1 | XP_016859013.1 | 37, 38 | 6228 | |||
GenBank mRNA | AB051484.2 | - | 1, 2 | 163 | ||
AB290165.1 | - | 3, 4 | 367 | |||
BC015442.1 | - | 2, 3 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
