siRNA Details
Gene Symbol: LRRC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ422148.1 | - | 2 | 1213 | ||
BC025744.1 | - | 3 | 1039 | |||
BC053659.1 | - | 3 | 1052 | |||
BC071640.1 | - | 2 | 897 | 5216040 | ||
BC093842.1 | - | 2 | 938 | 7939687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
