siRNA Details
Gene Symbol: GNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000172.3 | NP_000163.2 | 8 | 1063 | ||
NM_144499.2 | NP_653082.1 | 8 | 1063 | |||
GenBank mRNA | AF493908.1 | - | 8 | 947 | ||
BC095505.1 | - | 8 | 950 | 6519160 | ||
X15088.1 | - | 8 | 1050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
