siRNA Details
Gene Symbol: ZNF688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024683.1 | NP_001019854.1 | 3 | 428 | ||
NM_145271.3 | NP_660314.1 | 3 | 1116 | |||
XM_005255139.2 | XP_005255196.1 | 3 | 345 | |||
XM_005255140.2 | XP_005255197.1 | 3 | 965 | |||
GenBank mRNA | AK122680.1 | - | 3 | 1444 | ||
BC018997.2 | - | 3 | 395 | 4336167 | ||
BM011208.1 | - | 3 | 391 | |||
KU178870.1 | - | 3 | 338 | |||
KU178871.1 | - | 2 | 648 | |||
KU178872.1 | - | 2 | 224 |
