siRNA Details
Gene Symbol: TMEM217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286401.1 | NP_001273330.1 | 2, 3 | 1212 | ||
XM_011514370.2 | XP_011512672.1 | 1, 2 | 97 | |||
GenBank mRNA | BC026012.1 | - | 2, 3 | 651 | 5269554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
