siRNA Details
Gene Symbol: SSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018070.4 | NP_060540.2 | 5, 6 | 750 | ||
NM_145716.3 | NP_663768.1 | 5, 6 | 750 | |||
XM_017000897.1 | XP_016856386.1 | 5, 6 | 419 | |||
XM_017000898.1 | XP_016856387.1 | 2, 3 | 646 | |||
GenBank mRNA | AF500116.1 | - | 5, 6 | 578 | ||
AK001217.1 | - | 1, 2 | 98 | |||
AL519196.3 | - | 5, 6 | 725 | |||
BC003605.1 | - | 5, 6 | 389 | 3546126 | ||
BC066365.1 | - | 5, 6 | 494 | 6169677 | ||
DN602300.1 | - | 5, 6 | 750 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
