siRNA Details
Gene Symbol: ZNF688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024683.1 | NP_001019854.1 | 3 | 1047 | ||
NM_145271.3 | NP_660314.1 | 3 | 1735 | |||
XM_005255139.2 | XP_005255196.1 | 3 | 964 | |||
XM_005255140.2 | XP_005255197.1 | 3 | 1584 | |||
GenBank mRNA | AF085979.1 | - | 1 | 387 | ||
AK122680.1 | - | 3 | 2063 | |||
BC018997.2 | - | 3 | 1014 | 4336167 |
