siRNA Details
Gene Symbol: DHX57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198963.1 | NP_945314.1 | 13, 14 | 2649 | ||
XM_011533154.2 | XP_011531456.1 | 13, 14 | 2653 | |||
XM_011533155.2 | XP_011531457.1 | 13, 14 | 2651 | |||
XM_011533156.2 | XP_011531458.1 | 13, 14 | 2653 | |||
XM_011533157.1 | XP_011531459.1 | 13, 14 | 2653 | |||
GenBank mRNA | AB209249.1 | - | 8, 9 | 1136 | ||
AF283512.1 | - | 9, 10 | 1340 | |||
AK296739.1 | - | 13, 14 | 2688 | |||
AK307514.1 | - | 13, 14 | 2678 | |||
BC039270.1 | - | 12, 13 | 2479 | |||
BC053623.1 | - | 7, 8 | 950 | |||
BC065278.1 | - | 8, 9 | 1144 | |||
BC131534.1 | - | 13, 14 | 2621 | 40082400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
