siRNA Details
Gene Symbol: ZSCAN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514406.2 | XP_011512708.1 | 5 | 1584 | ||
XM_011514408.2 | XP_011512710.1 | 6 | 1771 | |||
XM_017010479.1 | XP_016865968.1 | 5 | 4800 | |||
GenBank mRNA | AK092117.1 | - | 6 | 1647 |
Gene Symbol: CYYR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135515.1 | 3 | 529 | |||
GenBank mRNA | AK125338.1 | - | 3 | 529 | ||
JF693915.1 | - | 3 | 366 |
Gene Symbol: OLFML2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000967.1 | XP_016856456.1 | 7 | 6559 |
Gene Symbol: SMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124545.1 | - | 1 | 637 |
Gene Symbol: TIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145702.2 | NP_663748.1 | 1 | 878 | ||
GenBank mRNA | AK056329.1 | - | 1 | 804 | ||
BC035143.1 | - | 1 | 883 | 5264460 | ||
BC063500.1 | - | 1 | 838 | 5248661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
