siRNA Details
Gene Symbol: HS6ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077188.1 | NP_001070656.1 | 3 | 1078 | ||
NM_147175.3 | NP_671704.3 | 3 | 1078 | |||
XM_005262490.3 | XP_005262547.1 | 2 | 978 | |||
XM_005262491.2 | XP_005262548.1 | 2 | 978 | |||
XM_011531406.1 | XP_011529708.1 | 2 | 307 | |||
XM_011531407.2 | XP_011529709.1 | 2 | 978 | |||
XM_011531408.2 | XP_011529710.1 | 2 | 978 | |||
XM_017029944.1 | XP_016885433.1 | 2 | 978 | |||
XM_017029945.1 | XP_016885434.1 | 2 | 252 | |||
XM_017029946.1 | XP_016885435.1 | 2 | 303 | |||
GenBank mRNA | AB067776.1 | - | 1 | 238 | ||
AB067777.1 | - | 1 | 225 | |||
AK027720.1 | - | 2 | 354 | |||
AK056706.1 | - | 3 | 1078 | |||
AK075402.1 | - | 2 | 257 | |||
AK295041.1 | - | 1 | 253 | |||
AK295118.1 | - | 1 | 218 | |||
AL049679.1 | - | 1 | 220 | |||
BC037325.2 | - | 1 | 137 | |||
BC094718.1 | - | 2 | 317 | |||
BC110620.1 | - | 2 | 917 | 40035499 | ||
BC110621.1 | - | 2 | 917 | 40035506 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001290468, NM_015819, NM_001077202, NM_001290467, BC063327, and others.
Gene Ontology Categories:
