siRNA Details
Gene Symbol: ZNF585B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152279.3 | NP_689492.3 | 5 | 698 | ||
GenBank mRNA | AK094989.1 | - | 1 | 700 | ||
AY147015.1 | - | 5 | 700 | |||
BC093625.1 | - | 4 | 540 | 7939470 | ||
BX440867.2 | - | 5 | 681 |
