siRNA Details
Gene Symbol: TMIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147196.2 | NP_671729.2 | 3 | 454 | ||
XM_006713097.3 | XP_006713160.1 | 3 | 1025 | |||
GenBank mRNA | AY081842.1 | - | 3 | 454 | ||
BC126258.1 | - | 3 | 417 | 8991974 | ||
BC126260.1 | - | 3 | 417 | 8991976 |
