siRNA Details
Gene Symbol: NRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006177.3 | NP_006168.1 | 3 | 1923 | ||
XM_005267708.4 | XP_005267765.1 | 4 | 2248 | |||
XM_005267709.3 | XP_005267766.1 | 4 | 2126 | |||
XM_005267710.3 | XP_005267767.1 | 3 | 1900 | |||
XM_011536801.2 | XP_011535103.2 | 3 | 2111 | |||
XM_011536802.1 | XP_011535104.1 | 4 | 3258 | |||
XM_011536804.2 | XP_011535106.1 | 4 | 2330 | |||
XM_011536805.2 | XP_011535107.1 | 3 | 1965 | |||
XM_011536806.2 | XP_011535108.2 | 3 | 1796 | |||
XM_017021350.1 | XP_016876839.1 | 3 | 1707 | |||
GenBank mRNA | AB593106.1 | - | 3 | 1934 | ||
M95925.1 | - | 3 | 1909 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
