siRNA Details
Gene Symbol: IZUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321449.1 | NP_001308378.1 | 3 | 436 | ||
NM_152358.2 | NP_689571.2 | 3 | 436 | |||
XM_011526436.2 | XP_011524738.1 | 3 | 376 | |||
XM_011526437.2 | XP_011524739.1 | 3 | 391 | |||
GenBank mRNA | AK131024.1 | - | 3 | 436 | ||
AY358196.1 | - | 3 | 350 | |||
BC029535.1 | - | 3 | 317 | |||
DB523757.2 | - | 3 | 208 |
