siRNA Details
Gene Symbol: ZNF684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152373.3 | NP_689586.3 | 5 | 1101 | ||
XM_011540671.2 | XP_011538973.1 | 6 | 1434 | |||
XM_011540672.2 | XP_011538974.1 | 6 | 1389 | |||
XM_011540673.2 | XP_011538975.1 | 5 | 1045 | |||
XM_017000290.1 | XP_016855779.1 | 6 | 2096 | |||
XM_017000291.1 | XP_016855780.1 | 4 | 998 | |||
XM_017000292.1 | XP_016855781.1 | 4 | 1186 | |||
GenBank mRNA | BC091518.1 | - | 5 | 1005 | 30367496 | |
BC111463.1 | - | 1 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
