siRNA Details
Gene Symbol: MUM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171020.1 | NP_001164491.1 | 4 | 749 | ||
NM_152423.4 | NP_689636.3 | 5 | 885 | |||
XM_005262077.2 | XP_005262134.1 | 4 | 666 | |||
XM_011530856.1 | XP_011529158.1 | 4 | 743 | |||
XM_011530857.1 | XP_011529159.1 | 4 | 712 | |||
XM_017029269.1 | XP_016884758.1 | 5 | 879 | |||
GenBank mRNA | AK056478.1 | - | 3 | 535 | ||
BC031229.1 | - | 4 | 749 | 5272099 | ||
BC109292.1 | - | 2 | 363 | |||
BC109293.1 | - | 2 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
