siRNA Details
Gene Symbol: CPT1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136052.2 | NP_001129524.1 | 8 | 1035 | ||
NM_001199752.2 | NP_001186681.1 | 8 | 1099 | |||
NM_001199753.1 | NP_001186682.1 | 8 | 1035 | |||
NM_152359.2 | NP_689572.1 | 7 | 966 | |||
NR_108072.1 | 9 | 1197 | ||||
XM_005258505.2 | XP_005258562.1 | 8 | 1072 | |||
XM_005258506.4 | XP_005258563.1 | 7 | 988 | |||
XM_006723009.3 | XP_006723072.1 | 6 | 705 | |||
XM_011526438.2 | XP_011524740.1 | 8 | 1128 | |||
XM_011526439.2 | XP_011524741.1 | 8 | 1192 | |||
XM_011526440.2 | XP_011524742.1 | 8 | 1044 | |||
XM_017026265.1 | XP_016881754.1 | 8 | 1091 | |||
XM_017026266.1 | XP_016881755.1 | 8 | 1045 | |||
XM_017026267.1 | XP_016881756.1 | 7 | 862 | |||
XM_017026268.1 | XP_016881757.1 | 6 | 705 | |||
XM_017026269.1 | XP_016881758.1 | 6 | 815 | |||
XM_017026270.1 | XP_016881759.1 | 6 | 583 | |||
XM_017026271.1 | XP_016881760.1 | 6 | 705 | |||
XM_017026272.1 | XP_016881761.1 | 7 | 899 | |||
XM_017026273.1 | XP_016881762.1 | 7 | 884 | |||
GenBank mRNA | AB208884.1 | - | 2 | 79 | ||
AF357970.1 | - | 7 | 924 | |||
AK096516.1 | - | 9 | 1193 | |||
AK289713.1 | - | 8 | 1091 | |||
AK290092.1 | - | 8 | 1007 | |||
AK299866.1 | - | 6 | 694 | |||
AK309087.1 | - | 7 | 925 | |||
BC029104.1 | - | 8 | 999 | 3872727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
