siRNA Details
Gene Symbol: FGFR1OP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007045.3 | NP_008976.1 | 7 | 631 | ||
GenBank mRNA | AK312791.1 | - | 7 | 542 | ||
Y18046.1 | - | 7 | 620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
