siRNA Details
Gene Symbol: TNFAIP8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167942.1 | NP_001161414.1 | 2 | 1673 | ||
NM_152362.2 | NP_689575.2 | 2 | 1645 | |||
XM_005259487.3 | XP_005259544.1 | 2 | 2360 | |||
XM_011527680.2 | XP_011525982.1 | 2 | 1651 | |||
GenBank mRNA | BC017672.1 | - | 2 | 1642 | 3885999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
