siRNA Details
Gene Symbol: EGFLAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205301.1 | NP_001192230.1 | 8 | 1210 | ||
NM_152403.3 | NP_689616.2 | 8 | 1210 | |||
NM_182798.2 | NP_877950.1 | 3 | 512 | |||
GenBank mRNA | AK096474.1 | - | 8 | 1188 | ||
AK097549.1 | - | 3 | 503 | |||
AK291602.1 | - | 9 | 1047 | |||
AL833357.1 | - | 4 | 366 | |||
BC063822.1 | - | 8 | 1051 | 6187990 | ||
BX647551.1 | - | 8 | 1060 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
