siRNA Details
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018071.3 | NP_001018081.3 | 29 | 4787 | ||
NM_001042512.2 | NP_001035977.2 | 6 | 1655 | |||
NM_001318191.1 | NP_001305120.1 | 27 | 4428 | |||
XM_017015744.1 | XP_016871233.1 | 6 | 1471 | |||
GenBank mRNA | AY491519.1 | - | 6 | 1654 | ||
BC031614.1 | - | 8 | 1650 | |||
BC073954.1 | - | 6 | 1636 | |||
CR749241.1 | - | 29 | 4759 |
Gene Symbol: FRMPD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033172.1 | 6 | 1709 | |||
GenBank mRNA | BC071635.1 | - | 6 | 1621 | ||
BC072395.1 | - | 6 | 1635 | |||
BC110798.1 | - | 6 | 1632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
