siRNA Details
Gene Symbol: LYSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057570.1 | - | 1 | 176 | ||
BC084545.1 | - | 3 | 609 | 4590701 | ||
BG420645.1 | - | 3 | 597 | |||
HY131530.1 | - | 3 | 378 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
