siRNA Details
Gene Symbol: ERN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI791744.1 | - | 1 | 171 | ||
AI792619.1 | - | 1 | 422 | |||
AK055561.1 | - | 1 | 1779 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
